Endurance Hi-Vis earmuffs, PS47
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PS47ORR PS47YER
PRTWPS47ORREA 18 EUR
PRTWPS47ORR PRTWPS47YER
Endurance Hi-Vis earmuffs, PS47
Oorkappen Oorkappen met Sound Management
These Easy to clip-on hard hat mounting earmuffs applies the latest standards to deliver the highest level of protection. Comfortable and lightweight, ensure a pleasant wear, even for long periods of use.
  • Hi-Visibility colour for visibility in daylight
  • Mad of ABS, polystyrene
  • Can be adjusted into the standoff position when not in use
  • Compatible with PS55, PW98, PS53, PV60, PV64, PV50, PV54, PS50 hard hats
  • Excellent attenuation rate
These hearing protection range includes PPE with different level sound attenuation, to be adapted to various working environments giving the correct protection from dangerous noise, without isolating the workers. These updated range of earplugs has best performance level.
Certificaten: AS/NZS 1270 (SLC(80) 22dB - Class 4); EN 352-3; ANSI S3.19 (NRR 24dB).
Verpakking: Retail bag which aids presentation for retail sales; Individually packed for vending machines.