Flame resistant light weight overalls, FR28
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
FR28RBRL FR28RBRM FR28RBRS FR28RBRXL FR28RBRXXL FR28RBRXXXL FR28RERL FR28RERM FR28RERS FR28RERXL FR28RERXXL FR28RERXXXL FR28NAR4XL FR28NAR5XL FR28NARL FR28NARM FR28NARS FR28NARXL FR28NARXS FR28NARXXL FR28NARXXXL FR28GRRL FR28GRRM FR28GRRS FR28GRRXL FR28GRRXXL
PRTWFR28RBRLEA 129 EUR
PRTWFR28RBRL PRTWFR28RBRM PRTWFR28RBRS PRTWFR28RBRXL PRTWFR28RBRXXL PRTWFR28RBRXXXL PRTWFR28RERL PRTWFR28RERM PRTWFR28RERS PRTWFR28RERXL PRTWFR28RERXXL PRTWFR28RERXXXL PRTWFR28NAR4XL PRTWFR28NAR5XL PRTWFR28NARL PRTWFR28NARM PRTWFR28NARS PRTWFR28NARXL PRTWFR28NARXS PRTWFR28NARXXL PRTWFR28NARXXXL PRTWFR28GRRL PRTWFR28GRRM PRTWFR28GRRS PRTWFR28GRRXL PRTWFR28GRRXXL
Flame resistant light weight overalls, FR28
Werkkleding
This anti-static overall is made of the innovative flame-resistant Bizflame Plus™ fabric. Sew on flame resistant industrial wash tape.
  • Hook and loop cuffs for a secure fit
  • Radio loops for easy clipping of a radio
  • 10 pockets for ample storage
  • Knee pad pockets designed with an innovative two tier adjustable system
Concealed two-way front brass zip and two front slant chest pockets with concealed brass zips. Class 2 welding protection and also protection against radiant, convective and contact heat.
Weight: 280 g