Cleanroom face masks, Auckland, P410
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
P410BLUL P410BLUM P410BLUS
PRTWP410BLULEA 12.6 EUR
PRTWP410BLUL PRTWP410BLUM PRTWP410BLUS
Cleanroom face masks, Auckland, P410
Maskers Cleanroom Maskers
Economical half mask with low profile design providing a large field of vision.
  • The soft adjustable strap ensures perfect fit to wide face contours
  • Equipped with a dual (screw in) filter for enhanced breathing comfort
  • Sweat port on the face piece for hot and humid environments
Manufactured from a durable yet soft non allergic TPE material which provides an excellent fit.
Certificaten: EN 140
Leveringsinformatie: Filters sold separately.