Heat resistant gloves, Duo-Therm A185
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A185Y4RL A185Y4RXL A185O4RXXL A185Y4RXXL A185Y4RM A185O4RL A185O4RXL A185O4RM
PRTWA185Y4RLEA 47 EUR
PRTWA185Y4RL PRTWA185Y4RXL PRTWA185O4RXXL PRTWA185Y4RXXL PRTWA185Y4RM PRTWA185O4RL PRTWA185O4RXL PRTWA185O4RM
Heat resistant gloves, Duo-Therm A185
Handschoenen
The A185 uses the latest techniques to produce an excellent and durable glove.
  • Thermal liner provides added warmth for the hand
  • Double dipping for maximum liquid repellency
  • 7 gauge liner for dexterity
  • Low linting construction for minimal contamination
An extra layer of wrinkled latex is added to the 'high-wearing' fingertip areas of the glove only, which improves grip and durability, while reducing hand fatigue.
Certificaten: EN 388:2016 (1131X), EN 511 (X1X), AS/NZS 2161 .3, AS/NZS 2161 .2, AS/NZS 2161 .5