Chemical resistant gloves, A813
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A813GNRL A813GNRM A813GNRXL A813GNRXXL
PRTWA813GNRLEA 128 EUR
PRTWA813GNRL PRTWA813GNRM PRTWA813GNRXL PRTWA813GNRXXL
Chemical resistant gloves, A813
Handschoenen
The extended length (480 mm) nitrile gauntlet offers excellent protection against many solvents.
  • Suitable for use in chemical, oil and food industries
  • Latex free (100%)
  • Silicone free - Ideal for manufacturing, paint applications, electronics and glass handling where silicone is problematic
  • Suited for food processing plants. resistant to greases, animal fats and oils
  • Retail bag which aids presentation for retail sales
  • Approved for safe food handling
Deigned with an anti-slip pattern, offers good grip in wet and dry conditions. The 0.55 mm thicknessMore Product Information