Thermal soft grip gloves, A143
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A143Y8RL A143Y8RM A143Y8RXL
PRTWA143Y8RLEA 3.3 EUR
PRTWA143Y8RL PRTWA143Y8RM PRTWA143Y8RXL
Thermal soft grip gloves, A143
Handschoenen
Great for cold environments, this glove is dipped with foamed latex offering extreme breathability.
  • Warm 10 gauge acrylic liner for cold protection
  • Latex foam coating for excellent grip in wet and dry conditions
  • Palm dipped to increase dexterity and ventilation
  • Breathable seamless liner
  • Ergonomically designed for comfort and ease of use
Provides superior grip performance in wet or dry conditions.
Certificaten: EN 388:2016 (2142X), EN 511 (X1X)