Sleeve protectors, cut resistant, A690
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A690GRR A690YER
PRTWA690GRREA 90.5 EUR
PRTWA690GRR PRTWA690YER
Sleeve protectors, cut resistant, A690
Werkkleding
These Cut level D resistance protective tubular sleeves offer cut and heat resistance for wrist and arm area.
  • Thumb hole for secure fit
  • Machine washable
  • Sold singly
  • Retail bag which aids presentation for retail sales
  • Seamless 13 gauge liner
Certificaten: AS/NZS 2161 .2; AS/NZS 2161 .3; AS/NZS 2161 .4; ANSI/ISEA 105: 2016 CUT Level A4; EN388:2016 +A1:2018 - 4.X.4.X.D.; EN 407 X.1.X.X.X.X.; EN 420, CE Food Safe.