Work overalls with bib and braces, PW346
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PW346ZBRL PW346ZBRM PW346ZBRS PW346ZBRXL PW346ZBRXXL PW346ZBRXXXL PW346BKRL PW346BKRM PW346BKRS PW346BKRXL PW346BKRXXL PW346BKRXXXL
PRTWPW346ZBRLEA 65.3 EUR
PRTWPW346ZBRL PRTWPW346ZBRM PRTWPW346ZBRS PRTWPW346ZBRXL PRTWPW346ZBRXXL PRTWPW346ZBRXXXL PRTWPW346BKRL PRTWPW346BKRM PRTWPW346BKRS PRTWPW346BKRXL PRTWPW346BKRXXL PRTWPW346BKRXXXL
Work overalls with bib and braces, PW346
Werkkleding
These work bib and brace showcases the best design, construction and performance. It offers a superb range of pockets including a double rule pocket, pre-shaped top loading knee pad pockets and an easy access thigh pocket.
  • Durable polycotton fabric for high performance and maximum wearer comfort
  • ID card holder for ID card display
  • Easy access cargo pocket
  • Hammer loop for secure storage of work tools
  • Double rule pocket
Contrast fabric: 100% polyester oxford 600D 210 g.
Shell fabric: Kingsmill 300 g.
Certificaten: EN 14404:2004+A1:2010 Type 2
Order Now
Specificaties
Model PW346
Environmentally Preferable
Kledingstuk toepassing General Purpose,Industrial
Risk area
Geslacht Uniseks
Sleeve or leg length Chest long
Stof Polyester