Chemical resistant gloves, Nitrosafe A810
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A810GNRL A810GNRM A810GNRS A810GNRXL A810GNRXXL
PRTWA810GNRLEA 36.4 EUR
PRTWA810GNRL PRTWA810GNRM PRTWA810GNRS PRTWA810GNRXL PRTWA810GNRXXL
Chemical resistant gloves, Nitrosafe A810
Handschoenen
Chemical resistant gauntlet with textured pattern for enhanced grip.
  • Textured pattern for enhanced grip
  • Fully coated for maximum liquid protection
  • Flock lined for added comfort
For use in the chemical, oil and food industries.
Certificaten: EN ISO 374-1:2016 Type A (AJKLMNOT), EN ISO 374-5 Micro Organisms, EN 388:2016 (3101X); AS/NZS 2161 .2, AS/NZS 2161 .3, AS/NZS 2161 .10.1