General purpose gloves, Dermi Pro Glove, AP30
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AP30O8RL AP30O8RM AP30O8RS AP30O8RXL AP30O8RXXL
PRTWAP30O8RLEA 52.8 EUR
PRTWAP30O8RL PRTWAP30O8RM PRTWAP30O8RS PRTWAP30O8RXL PRTWAP30O8RXXL
General purpose gloves, Dermi Pro Glove, AP30
Handschoenen
These fully dipped nitrile coated gloves with foam nitrile coating in the palm area provides comfort and touch sensitivity. An elastic knit cuff holds it firmly on the arm for enhanced fit. The nitrile coating offers excellent liquid barrier while the superior foam dip ensures excellent grip even in oily environments.
  • Super abrasion and tear resistance
  • Fully coated for maximum liquid protection
  • Nitrile foam coating for excellent grip in wet and dry conditions
  • Ergonomic design to reduce hand fatigue
  • 13 gauge liner for a perfect fit
  • Retail tag which aids