LH software, TRILUTION®
Leverancier: Gilson
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
GILI21063024EA 7041 EUR
GILI21063024
LH software, TRILUTION®
Software Laboratoriumsoftware
TRILUTION® LH provides a solution for customers using Windows 10 and is a comprehensive software package for seamless automation of all liquid handling and solid phase extraction (SPE) methods. With its intuitive interface, simple drag-and-drop method creation, and application simulation, it combines flexibility and ease of use.
  • Simple drag-and-drop method creation
  • Extensive flexibility in methods and tasks through use of variables and operators
  • Integrated customisation to create racks and tasks
  • Graphical application simulation to ensure the functionality and reliability of the application prior to use with precious samples
  • Full control of all SPE Key Parameters
  • Secured users access level
  • Multiple exportable reports
Five E's software for unmatched user experience:
Efficient: The application builder gives you access to everything you need to set up, run, and monitor your application from a single screen
Effective: Custom task and rack builders allow low-level access to customise the software to fit your needs
Engaging: Modern look and graphical task makes method development enjoyable
Error tolerant: The simulation helps prevent user errors, estimates run times, and ensures functionality and reliability of the application prior to use with precious samples
Easy to learn: Drag-and-drop functionality and a large selection of ready-to-use tasks makes method creation quick and easy
All-in-One software for optimised productivity:
TRILUTION software allows for multiple applications ranging from basic liquid handling (dilution, kinetic reaction) to sample preparation (filtration, liquid-liquid extraction (LLE), or solid phase extraction (SPE)) on the same liquid handler.