Cut resistant gloves, AP50
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AP50B8RXL AP50B8RXXL AP50B8RL AP50B8RS AP50B8RM
PRTWAP50B8RXLEA 130 EUR
PRTWAP50B8RXL PRTWAP50B8RXXL PRTWAP50B8RL PRTWAP50B8RS PRTWAP50B8RM
Cut resistant gloves, AP50
Handschoenen
The fully coated Aqua Cut Pro glove offers level D cut resistance.
  • Sandy finish for exceptional grip in water, grease or oil
  • Protects against cuts and abrasion
  • Prevents grease, oil and water penetration
  • Seamless 13 gauge liner
  • Retail bag which aids presentation for retail sales
  • ANSI Cut Level A4
The first coat is a smooth nitrile and the second coat is foam nitrile, providing a secure grip and a barrier against contaminants. Elastic wrist cuff provides a secure fit.
Certificaten: AS/NZS 2161 .2, AS/NZS 2161 .3, ANSI/ISEA 105 - 2016 CUT Level (A4), EN 420, EN 388:2016 - (4.X.4.4.D)