Safety boots, wellingtons, Portwest™ Pro FD95
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
FD95YER39 FD95YER40 FD95YER48 FD95YER41 FD95YER42 FD95YER44 FD95YER45 FD95YER46 FD95YER47 FD95YER43
PRTWFD95YER39EA 86.3 EUR
PRTWFD95YER39 PRTWFD95YER40 PRTWFD95YER48 PRTWFD95YER41 PRTWFD95YER42 PRTWFD95YER44 PRTWFD95YER45 PRTWFD95YER46 PRTWFD95YER47 PRTWFD95YER43
Safety boots, wellingtons, Portwest™ Pro FD95
Schoenen en laarzen
Antistatic boots made from more flexible and longer lasting than PVC or Rubber, due to its air bubble structure which provides air circulation. PU keeps feet hot in winter and cool in summer.
  • Protective steel toecap and steel midsole
  • Energy absorbing seat region
  • 100% waterproof keeping feet warm and dry
  • Slip resistant outsole to prevent slips and trips on ceramic and steel surfaces
  • Fuel and oil resistant outsole
  • Cold insulation
  • Puncture resistant non-metallic midsole (PR)
This range is designed to protect your feet in the toughest environments.
Certificaten: EN ISO 20345:2011, S5, SRC.