Corporate traffic jackets, S437
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
S437NARM S437NARS S437NARXXL S437NARXXXL S437NARL S437NARXL
PRTWS437NARMEA 63 EUR
PRTWS437NARM PRTWS437NARS PRTWS437NARXXL PRTWS437NARXXXL PRTWS437NARL PRTWS437NARXL
Corporate traffic jackets, S437
Werkkleding
These traffic jackets are designed to be worn in foul weather conditions, and offers excellent waterproof protection.
  • Waterproof keeping the wearer dry and protected from the elements
  • Taped seams to provide additional protection
  • Fully lined and padded to trap the heat and increase warmth
  • Storm flap front to protect against the elements
  • Two-way zip for quick and easy access
  • Four pockets for ample storage
  • Multiple utility pockets providing ample storage
  • Phone pocket
  • Pack away hood for added functionality
  • Print access for corporate branding
  • 300D Oxford Weave with a stain resistant finish, PU Coated
Weight (polyester lining fabric): 60 g/m²
Weight (polyester filling fabric): 170 g/m²
Weight (Shell fabric): 190 g/m²
Certificaten: CE certified, EN 343:2019 Class 3:1 X WP 15,000 mm, EN 342 0,349 m². K/W (B), 2, X
Opgelet: Wash at or before 40°C, Do not bleach, trumble dry, dry clean, iron and wring.