Bib and brace trousers, FR37, Bizflame™
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
FR37NARL FR37NARM FR37NARS FR37NARXL FR37NARXXL FR37NARXXXL
PRTWFR37NARLEA 77.8 EUR
PRTWFR37NARL PRTWFR37NARM PRTWFR37NARS PRTWFR37NARXL PRTWFR37NARXXL PRTWFR37NARXXXL
Bib and brace trousers, FR37, Bizflame™
Werkkleding
This classic bib provides complete lower body and chest protection. The large chest pocket is ideal for storing tools or personal belongings. Other handy features include a double rule pocket, a flapped back pocket and elasticated straps for comfort and ease of movement.
  • Guaranteed flame resistance for life of garment
  • Certified protection against molten metal splash
  • Protection against radiant, convective and contact heat and 50+ UPF rated fabric to block 98% of UV rays
  • Twin-stitched seams for extra durability
  • Durable, strong and long lasting brass zip and elasticated straps
  • Five pockets for ample storage and one chest pocket
Weight: 330 g/m²
Certificaten: EN 1149 -5, EN ISO 11612 (A1+A2, B1, C1, E3, F1), EN ISO 11611 Class 1 (A1+A2)