Occupational boots, pull-on, PVC Wellington 04, FW90
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
FW90GNR40 FW90GNR41 FW90GNR45 FW90GNR46 FW90GNR47 FW90GNR39 FW90GNR42 FW90GNR43 FW90GNR44
PRTWFW90GNR40EA 22.1 EUR
PRTWFW90GNR40 PRTWFW90GNR41 PRTWFW90GNR45 PRTWFW90GNR46 PRTWFW90GNR47 PRTWFW90GNR39 PRTWFW90GNR42 PRTWFW90GNR43 PRTWFW90GNR44
Occupational boots, pull-on, PVC Wellington 04, FW90
Schoenen en laarzen
These PVC wellington boots are suitable for workers in agriculture, fisheries, janitorial and light industry.
  • 100% waterproof keeping feet warm and dry
  • Slip resistant outsole to prevent slips and trips on ceramic and steel surfaces
  • Fuel and oil resistant outsole
Certificaten: EN ISO 20347