High visibility reversible bodywarmers, RT44
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
RT44ORRL RT44ORRM RT44ORRXL RT44ORRXXL RT44ORRXXXL RT44ORRS
PRTWRT44ORRLEA 59.8 EUR
PRTWRT44ORRL PRTWRT44ORRM PRTWRT44ORRXL PRTWRT44ORRXXL PRTWRT44ORRXXXL PRTWRT44ORRS
High visibility reversible bodywarmers, RT44
Werkkleding
High visibility reversible bodywarmer provides warmth and great flexibility. This unique design provides two deep pockets on both the hi-vis orange side and the reversed grey side.
  • Reversible garment
  • Texpel finish
  • Deep storage pockets
  • Quilt lined
These fabrics offer variations in weight and composition. The bright fluorescent capabilities of these fabrics combined with the garments retro reflective stripes provides the possible visibility in poor light or adverse weather conditions.
Shell fabric: 80% polyester, 20% cotton, 280 g
Lining fabric: 65% polyester, 35% cotton, 260 g
Filling fabric: 200 g
Certificaten: EN ISO 20471 Class 2, EN ISO 20471, EN 343, GO/RT 3279 Issue.