Safety boots, lace-up, Compositelite™ Traction, FD01
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
FD01BKR41 FD01BKR46 FD01BKR40 FD01BKR42 FD01BKR44 FD01BKR48 FD01BKR47 FD01BKR45 FD01BKR38 FD01BKR39 FD01BKR43
PRTWFD01BKR41EA 200 EUR
PRTWFD01BKR41 PRTWFD01BKR46 PRTWFD01BKR40 PRTWFD01BKR42 PRTWFD01BKR44 PRTWFD01BKR48 PRTWFD01BKR47 PRTWFD01BKR45 PRTWFD01BKR38 PRTWFD01BKR39 PRTWFD01BKR43
Safety boots, lace-up, Compositelite™ Traction, FD01
Schoenen en laarzen
Anti-static boots made from full-grain leather plus a dual-density PU/rubber sole, resistant to heat (300 °C).
  • Composite toecap for added protection
  • Pierce resistant composite midsole
  • Energy absorbing seat region
  • 100% waterproof membrane
  • Slip resistant outsole to prevent slips and trips on ceramic and steel surfaces
  • Fuel and oil resistant outsole
Combining comfort, ankle support, unrivalled anti slip properties and the finest materials and components, these boots are revelation in footwear construction. Lightweight construction with fastening zip, S3 water resistant leather and a waterproof and breathable membrane to keep feet dry in the harshest of conditions.
Certificaten: EN ISO 20345:2011, S3, HRO, SRC; ASTM F2413-11; EN 61340-4-3 Class 3.