High visibility T-shirts, S478
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
S478YER5XL S478YERL S478YERM S478YERS S478YERXL S478YERXS S478YERXXL S478YERXXXL S478YER4XL
PRTWS478YER5XLEA 16.4 EUR
PRTWS478YER5XL PRTWS478YERL PRTWS478YERM PRTWS478YERS PRTWS478YERXL PRTWS478YERXS PRTWS478YERXXL PRTWS478YERXXXL PRTWS478YER4XL
High visibility T-shirts, S478
Werkkleding
The lightweight sports-type fabric provides unbeatable wearer comfort and great ventilation on those hot summer days.
  • Silver reflective tape for increased visibility
  • 50 UPF rated fabric to block 98% of UV rays
  • Generous fit for wearer comfort
  • Crew neck
Breathable fabric to draw moisture away from the body keeping the wearer cool, dry and comfortable. Quality wicking fabric finish enhances fabric drying and aids stain removal.
Shell Fabric : 100% polyester, bird eye knit 150 g
Certificaten: S478 + TX71 certified to EN ISO 20471 Class 3 ensemble.