High visibility two-tone Polo shirts, S479
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
S479ONRXXXL S479YNRL S479YNRM S479YNRS S479YNRXL S479YNRXXL S479YNRXXXL S479ONRXL S479ONRS S479ONRXXL S479ONRL S479ONRM
PRTWS479ONRXXXLEA 16.8 EUR
PRTWS479ONRXXXL PRTWS479YNRL PRTWS479YNRM PRTWS479YNRS PRTWS479YNRXL PRTWS479YNRXXL PRTWS479YNRXXXL PRTWS479ONRXL PRTWS479ONRS PRTWS479ONRXXL PRTWS479ONRL PRTWS479ONRM
High visibility two-tone Polo shirts, S479
Werkkleding
Modern design, safety and wearer comfort are the main features of the S479. The two-tone contrast is perfect for warm, dirty environments when hi-vis protection is required.
  • Silver reflective tape for increased visibility
  • 100% polyester moisture wicking fabric
  • 50+ UPF rated fabric to block 98% of UV rays
  • Contrast panel for protection against dirt
  • Taped neck seam for extra comfort
  • Generous fit and side vents for added comfort
Machine wash warm/mild and line dry. Do not bleach, iron, or dry clean.
Shell fabric : 100% polyester, bird eye knit 175 g
Certificaten: ANSI/ISEA 107-2015 Type O Class 1. EN ISO 20471 Class 1.