Rain jackets, S555
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
S555BKRS S555BKRL S555BKRM S555BKRXXL S555BKRXXXL S555BKRXL
PRTWS555BKRSEA 74.7 EUR
PRTWS555BKRS PRTWS555BKRL PRTWS555BKRM PRTWS555BKRXXL PRTWS555BKRXXXL PRTWS555BKRXL
Rain jackets, S555
Werkkleding
These high performance Outcoach jackets offers maximum waterproof, windproof and breathable protection for year round performance.
  • Abrasion resistant Oxford polyester
  • Wind resistant to protect against wind chill
  • Lined for added warmth and comfort
  • Front zip opening for easy access and storm flap front to protect against the elements
  • Water resistant zips to prevent water penetration
  • Hood with peak and drawstrings and pack away hood for added functionality
  • Four pockets for ample storage
Using a highly durable outer fabric, with an air permeable mesh lining, this jacket ensures total comfort in foul conditions. High specification features, such as water resistant zippers, ergonomic cut, engineered storm hood and multiple pockets provide superb function. A sporty fit and low profile design ensure this is the jacket of choice for demanding conditions.
Shell fabric: 100% polyester, PU coated, 190 g
Lining fabric: body lining: 100% polyester mesh, 65 g; sleeve: 100% polyester, 190T Taffeta, 60 g
Certificaten: EN 343 Class 3:3