Jackets, bizflame pro, FR35
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
FR35NARL FR35NARXL FR35NARXXXL FR35NARM FR35NARXXL FR35NARS
PRTWFR35NARLEA 62.8 EUR
PRTWFR35NARL PRTWFR35NARXL PRTWFR35NARXXXL PRTWFR35NARM PRTWFR35NARXXL PRTWFR35NARS
Jackets, bizflame pro, FR35
Werkkleding
These Bizflame Pro jackets provides comfort and outstanding protection against radiant, convective, contact heat and molten metal splash. They are flame resistance and anti-static
  • Chest pocket with a concealed mobile phone section
  • Radio loop for easy clipping of a radio
  • Durable, strong and long lasting brass zip
  • Chest pockets with stud closure
  • Four pockets for ample storage
  • 50+ UPF rated fabric to block 98% of UV rays
Shell Fabric : Bizflame Pro 330 g
Certificaten: CE-CAT III, EN 1149 -5, EN ISO 11612 A1+A2, B1, C1, E3, F1, EN ISO 11611 Class 1 A1+A2