Safety shoes, lace-up, Steelite™ Ultra Mersey Trainer, FT50
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
FT50GRR36 FT50GRR37 FT50GRR38 FT50GRR39 FT50GRR40 FT50GRR41 FT50GRR42 FT50GRR43 FT50GRR44 FT50GRR45 FT50GRR46 FT50GRR47 FT50GRR48
PRTWFT50GRR36EA 53.6 EUR
PRTWFT50GRR36 PRTWFT50GRR37 PRTWFT50GRR38 PRTWFT50GRR39 PRTWFT50GRR40 PRTWFT50GRR41 PRTWFT50GRR42 PRTWFT50GRR43 PRTWFT50GRR44 PRTWFT50GRR45 PRTWFT50GRR46 PRTWFT50GRR47 PRTWFT50GRR48
Safety shoes, lace-up, Steelite™ Ultra Mersey Trainer, FT50
Schoenen en laarzen
High performance upper design with 3D mesh lining which provides maximum comfort and breathability. This trend setting safety footwear has been constructed with a dual density PU/PU outsole, providing superior grip and energy absorption. Designed with a cushioned footbed reducing foot fatigue.
  • Protective steel toecap
  • Anti-static footwear
  • Energy absorbing seat region
  • Fuel and oil resistant outsole
  • Dual density sole unit
  • Upper material: Cow suede leather
SRC enabled, slip resistant outsole to prevent slips and trips on ceramic and steel surfaces.
Certificaten: EN ISO 20345:2011; S1, SRC.
Verpakking: 10 per box.