Safety boots, lace-up, Steelite™ WORK Metatarsal, FW22
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
FW22BKR38 FW22BKR39 FW22BKR40 FW22BKR41 FW22BKR42 FW22BKR43 FW22BKR44 FW22BKR45 FW22BKR46 FW22BKR47
PRTWFW22BKR38EA 60.8 EUR
PRTWFW22BKR38 PRTWFW22BKR39 PRTWFW22BKR40 PRTWFW22BKR41 PRTWFW22BKR42 PRTWFW22BKR43 PRTWFW22BKR44 PRTWFW22BKR45 PRTWFW22BKR46 PRTWFW22BKR47
Safety boots, lace-up, Steelite™ WORK Metatarsal, FW22
Schoenen en laarzen
Antistatic safety shoes made from split leather plus a dual density PU/rubber sole, resistant to heat (300 °C).
  • Protective steel toecap
  • Steel midsole
  • Energy absorbing seat region
  • Slip resistant outsole to prevent slips and trips on ceramic and steel surfaces
  • Fuel and oil resistant outsole
  • Integrated superior metatarsal protection for heavy duty industries
Leather upper, with tough steel metatarsal cover offers supreme protection and comfort for the wearer. Heat resistant outsole with the highest slip resistance grade makes this boot suitable for wearMore Product Information