High visibility waistcoats, JN14
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
JN14YERS JN14YERM JN14YERL
PRTWJN14YERSEA 6 EUR
PRTWJN14YERS PRTWJN14YERM PRTWJN14YERL
High visibility waistcoats, JN14
Werkkleding
The simple front fastening allows children to put on and take off themselves.
  • Lightweight and comfortable
  • Reflective tape for increased visibility
  • Hook and loop closure for easy access
Ideal for school trips, social club outings and outdoor activities where high visibility protection is needed.
Shell fabric : 100% polyester 125 g
Certificaten: EN 1150