Anti impact grip gloves, A721
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A721Y1RS A721Y1RL A721Y1RXL A721Y1RM
PRTWA721Y1RSEA 229 EUR
PRTWA721Y1RS PRTWA721Y1RL PRTWA721Y1RXL PRTWA721Y1RM
Anti impact grip gloves, A721
Handschoenen
The new generation of working gloves offering maximum grip with maximum impact protection. Abrasion resistant nitrile foam coating provides exceptional grip for water, grease and oil contact.
  • Lightweight and breathable seamless liner
  • Hook & Loop strap for secure fit all day long
  • Highly durable and impact resistant
  • Nitrile foam coating for excellent grip in wet and dry conditions
  • 13 gauge liner for a perfect fit
  • Maximum impact protection using TPR pod technology
This collection of gloves provides vital protection against impact to the hand from multiple hazards.
Certificaten: EN 388:2016 (4241XP), AS/NZS 2161 .3, AS/NZS 2161 .2, ANSI/ISEA 105: 2016 CUT Level (A1), ANSI/ISEA 138 Level 2 – Pass