Rain trousers, Iona lite lined, S482
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
S482NARL S482NARM S482NARS S482NARXL S482NARXXL
PRTWS482NARLEA 45 EUR
PRTWS482NARL PRTWS482NARM PRTWS482NARS PRTWS482NARXL PRTWS482NARXXL
Rain trousers, Iona lite lined, S482
Werkkleding
Cleverly designed to keep the wearer visible, safe and dry in foul weather conditions, the S482 is extremely practical and waterproof. Reflective tape offers superb protection in low light conditions whilst the padding traps the heat to make you warm and comfortable.
  • Abrasion resistant, 100% polyester, 300D Oxford weave with a stain resistant finish, PU coated, 190 g
  • Taped seams to provide additional protection
  • Reflective tape for increased visibility
  • Fully lined and padded to trap the heat and increase warmth
  • Fully elasticated waistband for ultimate wearer comfort
  • Stud adjustable hems for a secure fit
  • Extremely water resistant fabric finish, water beads away from fabric surface
100% polyester, Lining fabric (60 g), filling fabric (170 g).
Tested to withstand the rigours of everyday wear, each garment has been designed and manufactured with care to ensure total satisfaction wear after wear.
Certificaten: EN 343:2019 Class 3:1 X (WP 15000 mm), EN 342 (0.349M². K/W (B), 2, X).
Verpakking: Do not wring.