Safety ankle boots, lace-up, Clyde, Portwest® Pro, FD10
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
FD10BKR42 FD10BKR38 FD10BKR39 FD10BKR40 FD10BKR41 FD10BKR43 FD10BKR44 FD10BKR45 FD10BKR46 FD10BKR47 FD10BKR48
PRTWFD10BKR42EA 101 EUR
PRTWFD10BKR42 PRTWFD10BKR38 PRTWFD10BKR39 PRTWFD10BKR40 PRTWFD10BKR41 PRTWFD10BKR43 PRTWFD10BKR44 PRTWFD10BKR45 PRTWFD10BKR46 PRTWFD10BKR47 PRTWFD10BKR48
Safety ankle boots, lace-up, Clyde, Portwest® Pro, FD10
Schoenen en laarzen
Anti-static safety ankle boots made from full grain leather plus a dual-density PU/rubber sole, resistant to heat (300 °C).
  • Protective steel toecap
  • Steel midsole
  • Energy absorbing seat region
  • Slip resistant outsole to prevent slips and trips on ceramic and steel surfaces
  • Fuel and oil resistant outsole
  • Protective TPU scuff cap for high impact and to increase longevity
  • Cold insulation
Moisture wicking inner lining provides reliable protection against cold, damp and heat. Innovative outsole flex zones increase flexibility and stability for excellent support and comfort with every step.
Certificaten: EN ISO 20345:2011; S3, SRC, HRO, CI HI FO.