Safety ankle boots, lace-up, Compositelite™ WORK, FC21
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
FC21BKR36 FC21BKR37 FC21BKR38 FC21BKR39 FC21BKR40 FC21BKR41 FC21BKR42 FC21BKR43 FC21BKR44 FC21BKR45 FC21BKR46 FC21BKR47 FC21BKR48
PRTWFC21BKR36EA 48.3 EUR
PRTWFC21BKR36 PRTWFC21BKR37 PRTWFC21BKR38 PRTWFC21BKR39 PRTWFC21BKR40 PRTWFC21BKR41 PRTWFC21BKR42 PRTWFC21BKR43 PRTWFC21BKR44 PRTWFC21BKR45 PRTWFC21BKR46 PRTWFC21BKR47 PRTWFC21BKR48
Safety ankle boots, lace-up, Compositelite™ WORK, FC21
Schoenen en laarzen
Lightweight, 100% metal-free leather safety boot made from split leather upper and a dual desity PU/PU outsole.
  • Composite toecap for added protection
  • Anti-static footwear
  • Energy absorbing seat region
  • Fuel and oil resistant outsole
  • Dual density sole unit
Top class design and engineering that offers stylish and durable protection. Non metallic safety components ideal for industrial wear with a strong and lightweight construction.
Colour: Black
Certificaten: EN ISO 20345 S1; SRC certified.
Order Now
Specificaties
Environmentally Preferable