Safety boots, Wellingtons S5 M, Portwest™ Pro, Metatarsal safety FD85
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
FD85BKR47 FD85BKR38 FD85BKR42 FD85BKR39 FD85BKR37 FD85BKR45 FD85BKR46 FD85BKR40 FD85BKR48 FD85BKR44 FD85BKR43 FD85BKR41
PRTWFD85BKR47EA 81.4 EUR
PRTWFD85BKR47 PRTWFD85BKR38 PRTWFD85BKR42 PRTWFD85BKR39 PRTWFD85BKR37 PRTWFD85BKR45 PRTWFD85BKR46 PRTWFD85BKR40 PRTWFD85BKR48 PRTWFD85BKR44 PRTWFD85BKR43 PRTWFD85BKR41
Safety boots, Wellingtons S5 M, Portwest™ Pro, Metatarsal safety FD85
Schoenen en laarzen
The Metatarsal safety Wellington offers complete foot protection. The innovative, non-metallic design allows flexibility and comfort to the wearer. Helps protect against hazards in heavy industrial work environments.
  • Composite toecap (200 J)
  • Pierce resistant composite midsole
  • Metatarsal protection
  • Anti-static footwear
  • Energy absorbing seat region
  • PVC/nitrile upper and outsole
Constructed to provide unbeatable comfort, support and flexibility, this range is designed to protect your feet in the toughest environments.
Certificaten: EN ISO 20345:2011 S5