Coats, Biztex™ SMS, type PB[6], ST31
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ST31WHRXL ST31WHRXXL ST31WHRL
PRTWST31WHRXLEA 150 EUR
PRTWST31WHRXL PRTWST31WHRXXL PRTWST31WHRL
Coats, Biztex™ SMS, type PB[6], ST31
Wegwerpkleding
These coats provide a fluid and particulate barrier to protect the worker without sacrificing mobility and comfort.
  • Stud front closing
Biztex™ SMS products are manufactured with an integrated three layer polypropylene construction: spunbond / meltblown / spunbond giving a breathable barrier.
Certificaten: EN 13034 Type 6 PB.