Centrifuge tubes, sterile, polypropylene, Corning®
Leverancier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
431123 430776
734-1880EA 296.97 EUR
734-1880 734-1863
Centrifuge tubes, sterile, polypropylene, Corning®
Buisjes Centrifugebuisjes
PP, with HDPE plug seal cap, sterile. Polyetherimide cushions.
  • Moulded graduations, conical bottom
  • Tubes are sterile and certified non pyrogenic
  • Support cushions must be used unless the rotor has appropriately shaped V-bottom holders
Ideal for applications requiring large volume centrifugation.
They are perfect for large volume centrifugation applications. They incorporate plug seal caps and have molded graduations.
Bestelinformatie: Non sterile cushion sold separately.
Verpakking: Bulk, 6/zak.