Leggings, Thermal baselayer, B131
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
B131BKRS B131BKRM B131BKRL B131BKRXL B131BKRXXL B131CHAS B131CHAM B131CHAL B131CHAXL B131CHAXXL B131CHAXXXL B131CHA4XL
PRTWB131BKRSEA 18.2 EUR
PRTWB131BKRS PRTWB131BKRM PRTWB131BKRL PRTWB131BKRXL PRTWB131BKRXXL PRTWB131CHAS PRTWB131CHAM PRTWB131CHAL PRTWB131CHAXL PRTWB131CHAXXL PRTWB131CHAXXXL PRTWB131CHA4XL
Leggings, Thermal baselayer, B131
Werkkleding
These smooth leggings have low profile seam construction for ultimate comfort.
  • Made from a 100% wicking polyester (140 g)
  • Retail box which aids presentation for retail sales
The material which draws moisture away from the body keeping the wearer cool, dry and comfortable under exerting conditions. The garments are suitable for all levels of activity and can be worn as an extra thermal layer for workwear in cold weather or as a technical sports garment for added comfort and enhanced performance.
Opgelet: Wash dark colours separately.