Accessories for ion selective electrodes, Orion™
Leverancier: Thermo Orion
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
948201 9700BNWP 9300BNWP
662-1544EA 184 EUR
662-1544 ORIO9700BNWP ORIO9300BNWP
Accessories for ion selective electrodes, Orion™
Elektroden ISE-elektroden
Accessories for solid state and plastic membrane ion selective electrodes.
  • Electrodes handles are used with plastic membrane combination or half-cell electrodes
  • Polishing strips help restore the performance of solid-state, half-cell, or combination ion selective electrodes