ESD safety shoes, slip-on, uvex 1 sport hygiene, S2, 6581
Leverancier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6581.7/35 6581.7/36 6581.7/37 6581.7/38 6581.7/39 6581.7/40 6581.7/41 6581.7/42 6581.7/43 6581.7/44 6581.7/45 6581.7/46 6581.7/47 6581.7/48 6581.7/49 6581.7/50 6581.7/51 6581.7/52 6581.8/35 6581.8/36 6581.8/37 6581.8/38 6581.8/39 6581.8/40 6581.8/41 6581.8/42 6581.8/43 6581.8/44 6581.8/45 6581.8/46 6581.8/47 6581.8/48 6581.8/49 6581.8/50 6581.8/51 6581.8/52 6581.9/35 6581.9/36 6581.9/37 6581.9/38 6581.9/39 6581.9/40 6581.9/41 6581.9/42 6581.9/43 6581.9/44 6581.9/45 6581.9/46 6581.9/47 6581.9/48 6581.9/49 6581.9/50 6581.9/51 6581.9/52
UVEX6581.7/35EA 83.3 EUR
UVEX6581.7/35 UVEX6581.7/36 UVEX6581.7/37 UVEX6581.7/38 UVEX6581.7/39 UVEX6581.7/40 UVEX6581.7/41 UVEX6581.7/42 UVEX6581.7/43 UVEX6581.7/44 UVEX6581.7/45 UVEX6581.7/46 UVEX6581.7/47 UVEX6581.7/48 UVEX6581.7/49 UVEX6581.7/50 UVEX6581.7/51 UVEX6581.7/52 UVEX6581.8/35 UVEX6581.8/36 UVEX6581.8/37 UVEX6581.8/38 UVEX6581.8/39 UVEX6581.8/40 UVEX6581.8/41 UVEX6581.8/42 UVEX6581.8/43 UVEX6581.8/44 UVEX6581.8/45 UVEX6581.8/46 UVEX6581.8/47 UVEX6581.8/48 UVEX6581.8/49 UVEX6581.8/50 UVEX6581.8/51 UVEX6581.8/52 UVEX6581.9/35 UVEX6581.9/36 UVEX6581.9/37 UVEX6581.9/38 UVEX6581.9/39 UVEX6581.9/40 UVEX6581.9/41 UVEX6581.9/42 UVEX6581.9/43 UVEX6581.9/44 UVEX6581.9/45 UVEX6581.9/46 UVEX6581.9/47 UVEX6581.9/48 UVEX6581.9/49 UVEX6581.9/50 UVEX6581.9/51 UVEX6581.9/52
ESD safety shoes, slip-on, uvex 1 sport hygiene, S2, 6581
Schoenen en laarzen
Modern, metal-free, ultra-lightweight and flexible S2 safety slip-on shoe.
  • Upper made from water repellent microfibre material that is easy to clean
  • Minimal number of seams to reduce contamination
  • Suitable for people allergic to chrome as it’s made from synthetic materials
  • Washable at 30 °C
  • All sole materials are free of silicones, plasticisers, and other paint wetting impairment substances
  • Meets the ESD specification with volume resistance less than 35 megaohms
  • Soft padding on collar and tongue
100% metal-free uvex xenova® toe cap, with a compact design, anatomical shape, good lateral stability, and no thermal conductivity. The ergonomically designed outsole is comprised of dual-density polyurethane with good shock absorption and excellent slip resistance.
A removable anti-static comfortable insole provides the foot with exceptional, full-surface shock absorption and ensures comfort in high-strain zones.
Certificaten: EN ISO 20345:2011 S2 SRC.