Cut resistant gloves, Razor - Lite, A630
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A630G7RM A630G7RL A630G7RXL
PRTWA630G7RMEA 198 EUR
PRTWA630G7RM PRTWA630G7RL PRTWA630G7RXL
Cut resistant gloves, Razor - Lite, A630
Handschoenen
Cut resistant level D gloves with reinforced leather palm for excellent heat protection, enhanced grip and prolonged glove life.
  • Tested for both cut and heat protection
  • Additional reinforced protection on palm and forefinger areas
  • Designed for tasks that require reinforced abrasion resistance
  • 13 gauge liner for a perfect fit
Ideal for glass handling and working with hot items up to 250 °C for short periods.
Certificaten: AS/NZS 2161 .2, AS/NZS 2161 .3, AS/NZS 2161 .4, ANSI/ISEA 105 - 2016 CUT Level (A4), EN 388:2016 (4X42D), EN 407 (X2XXXX); Cat. II