Sliding beam anchor, DBI-SALA, 3M™
Leverancier: 3M
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
DBI-SALA®
115-2596EA 0 EUR
115-2596
Sliding beam anchor, DBI-SALA, 3M™
Valbescherming Veiligheidslijnen
The new 3M™ DBI-SALA® Sliding Beam Anchor offers the best of both mobility and reliable fall protection security. Once secured to the beam using the redesigned adjustment buttons, simply connect a Self-Retracting Lifeline (SRL) or lanyard to the swivelling D-ring to begin work. The anchor point slides effortlessly behind the user.
  • Hands-free mobility
  • 360-degree swivelling D-ring optimizes mobility on the beam
  • Compact size and lightweight materials
  • Easy pad replacement
  • Snap-in design of wear pads allows them to replace quickly in the field
Corrosion-resistant construction meets CE standards for weather, environmental, and storage conditions. Visible adjustability dual ratcheting adjustment buttons are accessible above the beam, while locking mechanism is located on the side to reduce accidental engagement.
Certificaten: EN 795-2012 Class B