Self-retracting lifelines, Rebel™, Protecta®, 3M™
Leverancier: CAPITAL SAFETY
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3100420 3100423 3590498 3590520 3590521 3590545 3590610
CPSD3100420EA 0 EUR
CPSD3100420 CPSD3100423 115-2678 CPSD3590520 CPSD3590521 CPSD3590545 CPSD3590610
Self-retracting lifelines, Rebel™, Protecta®, 3M™
Valbescherming Veiligheidslijnen
These Protecta® Rebel™ Web self-retracting lifelines provide an economical fall protection solution without compromising on performance or safety. They are available in a range of lengths to meet the specific needs of jobsites. Lifelines are available in a super-tough nylon housing comprised of 25 mm polyester webbing, or in a durable, lightweight thermoplastic housing comprised of galvanised steel.
  • Swivelling top anchorage point
  • Self-locking snap hook on lifeline
  • Compact, lightweight design
  • Tough and impact-resistant stackable thermoplastic housing
  • Quick activating speed sensing brake system
  • High-capacity external energy absorber and impact indicator
  • Heat-shrunk PVC coating on the shock pack
The SRL’s lifeline will extend as the user moves away, and retract automatically enabling the user to move about within a recommended working area at normal speeds. Should a fall occur, a speed sensing brake system will activate stopping the fall and reducing the forces imposed on the user to safe levels. The Rebel™ features an integrated, ergonomic carrying handle for easy transportation and a top swivel connector for quick installation. The built-in fall indicator hook allows visual inspection. Available in a variety of lengths and lifeline material options, with a snap hook carabiner, there is a Rebel™ SRL to meet many different applications and industries.
The mounting bracket is designed to attach the Protecta® Rebel Retrieval Self-Retracting Lifeline to a Protecta® tripod AM100, specifically designed for confined space applications.
Certificaten: Certified to EN360.