VWR® Reagent Bottles with PP 28 Thread
Leverancier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
215-4403EA 78.7 EUR
215-4403 215-4404 215-4405 215-4406 215-4407 215-4408 215-4409 215-4410 215-4411 215-4412 215-4413 215-4414 215-4415 215-4416 215-4417 215-4418
VWR® Reagent Bottles with PP 28 Thread
Flessen Glazen flessen
Made from clear or amber soda-lime glass. Ideal for sampling, storage, transporting liquids, or other applications that do not involve high thermal stress.
  • Good chemical and physical properties
  • Clear or amber glass
  • Amber glass bottles ideal for use with light-sensitive liquids
Bottles can be sterilised but the temperature difference (Δ T) must not exceed 40 °C.
Bestelinformatie: Supplied without caps, screw caps must be ordered separately.