Electronic test system
Leverancier: 3M MEDICAL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4109 4308
MMME4109EA 503 EUR
MMME4109 MMME4308
Electronic test system
Sterilisatie-indicatoren
The Electronic Test System (ETS) is a stand alone test device for measurements of the physical parameters of a steam sterilisation cycle providing an accurate temperature, pressure and time measurement.
  • Service parts available to authorized entities only
  • Fully automatic with pass/fail Bowie-Dick type test result
  • Data download to PC software for capture and trending
With the use of the ETS software and the data-reader the data can be downloaded to the software. With the software the data can be analysed and stored in a digital logbook.
Segment: life sciences - sterilisation