Fraction collectors
Leverancier: GILSON LCMS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
171011 171011DV 171041
GILI171011EA 3910 EUR
GILI171011 GILI171011DV GILI171041
Fraction collectors
Fractieverzamelaars
Ideal solutions for small to large capacity collection. Gilson offers advanced solutions for fraction collection with a complete line of stand‑alone and software-controlled fraction collectors ranging from small footprint to large vessel capacity models. The built-in, easy-to-use keypad makes setup and operation simple and capable of interfacing with any HPLC system or control the system through your PC with TRILUTION® LC software.
  • Collect fractions by time, drop, or peak to suit all your LC applications
  • Choose the compact FC 203B for small-capacity collection or the FC 204 for large volumes and capacities
  • In peak mode, collectors monitor a detector signal and identify peaks using either an adaptive-slope algorithm or a specified millivolt threshold
  • Time: Up to 10 programmable time windows can be added for collecting only what you want, while discarding the column’s void volume, peaks of no interest, and equilibration volumes
  • 3-way diverter valve prevents contamination of collected fractions (optional)
  • Compact size fits in most fume hoods
FC203B and FC203B with valve: choose among peak, drop, or time collection options with the FC 203B fraction collector. In peak mode, the FC 203B monitors a detector signal and identifies peaks using either an adaptive-slope algorithm or a specified millivolt threshold. With up to ten programmable time windows to use, collect only what you want and discard the column void volume, peaks of no interest, and equilibration volumes. Multicolumn adapters are available for collecting up to 18 separate channels and an optional 3-way diverter valve prevents contamination of collected fractions.
FC204: Collect samples by peak, drop, or time using the FC 204 Fraction Collector. Add up to ten programmable time windows to collect only what you want, eliminating void volumes, equilibration volumes, and peaks of no interest. In peak mode, the collector monitors a detector signal, identifying peaks using an adaptive-slope algorithm or a specified millivolt threshold. The 3-way diverter valve prevents contamination of fractions. Multicolumn adapters allow for collection from up to 18 separate channels.