CPC systems
Leverancier: GILSON LCMS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
21141008 21141010 21141007
GILI21141008EA 122620 EUR
GILI21141008 GILI21141010 GILI21141007
CPC systems
Kolommen voor chromatografie
Gilson’s reusable, silica-free centrifugal partition chromatography (CPC) columns offer high injection capacities of milligram to multi-kilograms resulting in 95% recoveries with 99% purity or better.
  • Increased savings
  • Improved purification efficiency
  • Numerous column options
CPC columns use approximately five times less solvent than more traditional columns, saving you money on solvent and disposal costs. Likewise, the reusable columns eliminates column replacement and silica recycling costs. CPC columns offer high sample injection capacities fromMore Product Information