Safety funnels with lid, 'MARCO' V2.0, electrically conductive
Leverancier: S.C.A.T.
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
317637 317651 318960 318961 318962 318964 318980 318981 318983 318984 318985 318988 306980 318630
221-0296EA 259 EUR
221-0296 SCTV317651 221-0299 221-0300 221-0301 SCTV318964 SCTV318980 SCTV318981 221-0302 SCTV318984 SCTV318985 SCTV318988 SCTV306980 SCTV318630
Safety funnels with lid, 'MARCO' V2.0, electrically conductive
Sluitingen Veiligheidsdoppen en veiligheidsafvaldoppen
S.C.A.T. 'MARCO' V2.0 safety funnels can be closed safely and prevent danger of ignition.
  • 'MARCO', V2.0 safety lid funnel is a modern lid funnel made of high-quality HDPE with extremely high resistance to acids, alkalis and many chemicals
  • Integrated gripping edge on the funnel lid increases the grip without having to touch the possibly contoured lid
  • Innovative funnel lid with integrated sealing ring closes the funnel edge and prevents explosive or health-endangering gases escaping
  • Sturdy hinge guarantees the closing mechanism has a long service life
  • Rotating screw cap makes it easier to screw the lid funnel on and off the canister
  • With removable sieve for easy cleaning
  • The non-conductive version of the 'MARCO' V2.0 safety lid funnel is in line with the SCAT colour scheme with its blue and white colouring, the conductive version is black, this difference in colour helps differentiate the conductive from the non-conductive version
The innovative 'MARCO', V2.0 safety funnel is a high-quality lid funnel for the disposal of liquid waste. The lid, with an integrated rubber lip, seals the funnel and prevents the escape of toxic or explosive vapours. Numerous features guarantee safe handling. These electrically conductive versions has an earth connection to avoid ignition hazards. Available in different thread sizes for safe attachment to laboratory bottles, canisters or other containers.
Diameter: 140 mm
Leveringsinformatie: Supplied with grounding cable.