epT.I.P.S.® Racks - Pipette Tips
Leverancier: EPPENDORF
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0030075.226 0030075.234 0030075.242 0030075.250 0030075.269 0030075.277 0030075.285 0030075.293 0030075.307 0030075.315 0030071.646 0030071.638 0030071.654 0030071.662
613-6751EA 67.1 EUR
613-6751 613-6752 613-6753 613-6754 613-6755 613-6756 613-6757 613-6758 613-6759 613-6760 613-6934 613-6933 613-6935 613-6936
epT.I.P.S.® Racks - Pipette Tips
Pipettips
Eppendorf epT.I.P.S.® Racks feature high-precision pipette tips in 'Eppendorf Quality', 'PCR clean', 'Biopur®', and 'Sterile' purity grades. All tips are supplied in disposible racks designed for optimal organisation and user convenience in demanding laboratory environments.
  • Long, narrow tips reach the base of narrow and high vessels without touching the top of the vessel edge
  • Modern slim rack design with reclosable lid
  • Improved sustainability through 35% lower polypropylene consumption per rack
  • ACT® labeled by non-profit My Green Lab® to help make an