Western blot stripping buffer, restore PLUS™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
46430 46428
PIER46430EA 250 EUR
PIER46430 PIER46428
Western blot stripping buffer, restore PLUS™
Reagentia voor elektroforese Western Blotting Reagents
Some antibodies are difficult to remove from Western blots and require Longer incubation times or incubation temperatures greater than 22 °C. Restore™ PLUS Western Blot Stripping Buffer was developed to reduce incubation times while keeping incubations at room temperature. High-affinity antibodies can be quickly and effectively stripped from Western blots without removing transferred proteins, thereby allowing multiple reprobes of the target.
  • Ready and easy to use - no dilution necessary, no offensive odours and room temperature storage
  • Compatible with commonly used Western blotting reagents and other materials - use on nitrocellulose and PVDF membranes, stored wet or dry; works with blocking buffer, enzyme conjugate and chemiluminescent substrate of choice
  • Cost effective - saves valuable time and sample; strip blots effectively the first time
  • Robust, but gentle - transferred proteins remain viable; strip the same blot up to five times
  • Flexible - strip and re-probe to optimise antibody concentrations; strip and re-probe for new antigen of interest
Order Now
Specificaties
Environmentally Preferable