Film, Amersham Hyperfilm™ ECL
Leverancier: Cytiva
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
28-9068-36 28-9068-37 28-9068-35 28-9068-39 28-9068-38
28-9068-36EA 317 EUR
28-9068-36 28-9068-37 28-9068-35 28-9068-39 28-9068-38
Film, Amersham Hyperfilm™ ECL
Autoradiografiefilms Folies
Amersham™ Hyperfilm™ ECL™ is constructed from a clear base coated on both sides with a photographic emulsion that is protected by an anti-scratch layer. The anti-scratch coating reduces the risk of visible artefacts (e.g., plastic wrap marks) and the clear colourless base results in easy reading of images without compromising on sensitivity.
  • High sensitivity for low concentration of nucleic acid and protein targets
  • Optimised for use in all blue and green light chemiluminescence systems
  • Clear background for excellent contrast and improved band resolution