Standard vest style fall arrest harness, Protecta®, 3M™
Leverancier: CAPITAL SAFETY
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1161609 1161610 1161611 1161615 1161616 1161617 1161642 1161643 1161644
CPSD1161609EA 0 EUR
CPSD1161609 CPSD1161610 115-2663 CPSD1161615 CPSD1161616 CPSD1161617 CPSD1161642 115-2664 CPSD1161644
Standard vest style fall arrest harness, Protecta®, 3M™
Valbescherming Valharnassen
Protecta® Standard Vest Style Fall Arrest Harness is a sleek and modern safety harness with upgraded features. It has water and stain resistant webbing which keeps it clean and dry, and protected labels that stay readable in the long term for simple and easy inspection.
  • Stain and water resistant webbing keeps the harness cleaner and dryer
  • Steel, zinc-plated black electro-deposition paint D-rings
  • Conveniently placed front D-ring helps users stay balanced and centred
  • Auto-resetting lanyard keeper provides easy parking and release of snap hooks or carabiners
  • Impact indicators make it simple to inspect the harness for damage and help ensure it’s safe to use
  • Easy and clear identification of fall arrest anchorage points
  • No loose web ends help users stay comfortable and tangle-free
For reliable protection, comfort, and convenience in a variety of styles and configurations, use the Protecta® Fall Arrest Harness. This standard vest style fall arrest harness has enhanced features including water and stain resistant webbing and auto-resetting lanyard keepers. No loose web ends help users stay comfortable and tangle-free. Tools can be secured while working at heights, to keep people and property safe anywhere below the worksite.
The standard vest style fall arrest harness with horizontal leg straps features articulated rings on the horizontal leg straps that allow the upper and lower body to twist, providing greater freedom of movement.
Impact indicators make it simple to check the harness for damage and help ensure it’s safe to use. Covered identification labels help protect important information for the life of the harness.