Examination gloves, nitrile, Gen-X®, smart glove
Leverancier: SMART GLOVE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PS047B-L PS047B-M PS047B-XL PS047B-XS
112-6133EA 0 EUR
112-6133 112-6134 112-6136 112-6137
Examination gloves, nitrile, Gen-X®, smart glove
Handschoenen
These non sterile examination-grade gloves are made of nitrile.
  • Ambidextrous
  • Fully palm textured to provide a secure grip
  • Extra-soft and comfortable
  • Powder-free
  • Beaded cuffs allow easy donning
  • 100% latex-free to eliminate adverse reactions associated with natural rubber latex proteins
Ideal for use in industrial and laboratory applications.
Certificaten: EN ISO 374-1, EN 374-2, EN 374-4, EN374-5, EN 16523-1; Cat. III; AQL 1,5
Verpakking: 100 gloves per box.