Safety canisters, electrically conducting
Leverancier: S.C.A.T.
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
107953 108027 108042 108043 107940 107740 108420 108192 108193 108421 108214 108216 108215 108217 108317 108331
590-0650EA 65.3 EUR
590-0650 590-0717 590-0732 590-0733 590-0639 SCTV107740 590-0032 SCTV108192 SCTV108193 SCTV108421 SCTV108214 SCTV108216 SCTV108215 SCTV108217 SCTV108317 SCTV108331
Safety canisters, electrically conducting
Veiligheidscontainers
SCAT canisters made of high-quality electrically conductive PE-HD-el material to prevent flying sparks caused by electrical discharge when disposing of flammable or explosive laboratory liquids.
  • Made of the very robust and electronically dissipative HDPE, which is extremely resistant to all solvents commonly used in laboratories
  • Filling quantities from 10 L for the use with HPLC systems up to massive 60 L drums for the collection of large quantities of containers in e.g. professional SymLine disposal systems
  • Thread types S50, S60/61, S70/71 and S90 are
More Product Information
Order Now
Specificaties
Environmentally Preferable