Space-saving canisters
Leverancier: S.C.A.T.
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
107998 108945
590-0690EA 69.1 EUR
590-0690 590-0795
Space-saving canisters
Veiligheidscontainers
SCAT space-saving canisters for use where space is limited in the laboratory, e.g. next to the HPLC system, in narrow spaces or safety cabinets. SCAT space-saving canisters offer the same high safety standards as conventional SCAT canisters.
  • Made of polypropylene (PP) with S50 thread
  • The capacity is standardised 5 L
  • Available as standard without float or optionally with a float for level control
SCAT space-saving canisters offer a clever alternative to conventional SCAT canisters by requiring only a fraction of the floor space for storage, without compromising on capacity.